Learn More
Medchemexpress LLC HY-100388 100mg Medchemexpress, SHP099 CAS:1801747-42-1 Purity:98%
SDP
Catalog No. 501636715
Encompass_Preferred
Medchemexpress,HY-100388 100mg SHP099 CAS:1801747-42-1 Formula:C16H19Cl2N5 SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC50 of 70 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6715 Supplier Medchemexpress LLC Supplier No. HY100388100MG
Add to Cart
Add to Cart
Medchemexpress,HY-100388 100mg SHP099 CAS:1801747-42-1 Formula:C16H19Cl2N5 SHP099 is a potent, selective, orally available SHP2 inhibitor with an IC50 of 70 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.