Learn More
Selleck Chemical LLC SHP099 HCl 5mg 2200214-93-1
SDP
Catalog No. 501365885
Encompass_Preferred
SHP099 is a highly potent, selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50?value of?0.071??M and shows no activity against SHP1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5885 Supplier Selleck Chemical LLC Supplier No. S82785MG
SHP099 is a highly potent, selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50?value of?0.071??M and shows no activity against SHP1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.