Learn More
Selleck Chemical LLC SHP099 HCl
SDP
Catalog No. 5000001042
Encompass_Preferred
SHP099 is a highly potent selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50 value of 0 071 M and shows no activity against SHP1
Catalog No. 50-000-01042 Supplier Selleck Chemical LLC Supplier No. S8278100MG
Add to Cart
Add to Cart
SHP099 is a highly potent selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50 value of 0 071 M and shows no activity against SHP1