Learn More
Medchemexpress LLC HY-100388A 100mg Medchemexpress, SHP099 (hydrochloride) CAS:1801747-11-4 Purity:98%
SDP
Catalog No. 501636716
Encompass_Preferred
Medchemexpress,HY-100388A 100mg SHP099 (hydrochloride) CAS:1801747-11-4 Formula:C16H20Cl3N5 SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6716 Supplier Medchemexpress LLC Supplier No. HY100388A100MG
Medchemexpress,HY-100388A 100mg SHP099 (hydrochloride) CAS:1801747-11-4 Formula:C16H20Cl3N5 SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.