Learn More
Sino Biological SHP-1/PTPN6 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 108192T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SHP-1/PTPN6, and purified by antigen affinity chromatography.
Catalog No. 50-224-8561