Learn More
Medchemexpress LLC SHP2-IN-6 hydrochloride | 2169223-49-6 | 98.9% | 25 MG
SDP
Catalog No. 5000342451
Encompass_Preferred
JAB-3068 hydrochloride is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM. This product is intended for research use only and is not for sale to patients.
  • Potent SHP2 inhibitor with an IC50 of 25.8 nM
  • Inhibits KYSE-520 cell proliferation with an IC50 of 2.17 μM
  • Extracted from patent WO2017211303A1, compound 7
  • For research use only
Catalog No. 50-003-42451 Supplier Medchemexpress LLC Supplier No. 25MGHY131132A
JAB-3068 hydrochloride is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM. This product is intended for research use only and is not for sale to patients.
  • Potent SHP2 inhibitor with an IC50 of 25.8 nM
  • Inhibits KYSE-520 cell proliferation with an IC50 of 2.17 μM
  • Extracted from patent WO2017211303A1, compound 7
  • For research use only