Learn More
Medchemexpress LLC SHP394 | 100MG
SDP
Catalog No. 5000229044
Encompass_Preferred
SHP394 is an orally active selective and allosteric inhibitor of SHP2 with an IC50 of 23 nM[1]
Catalog No. 50-002-29044 Supplier Medchemexpress LLC Supplier No. HY114397100MG
SHP394 is an orally active selective and allosteric inhibitor of SHP2 with an IC50 of 23 nM[1]