Learn More
Aviva Systems Biology Corp Shpk Antibody - C-terminal region (ARP54926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Shpk. It was validated on Western Blot. . Alternate names: 4930431K22Rik, AI194947, AW260459, CarklApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-978