Learn More
MilliporeSigma SHR OPTICAP XL600 3/PK
Supplier:  MilliporeSigma KVEPS006HB3
Encompass
Opticap Sterile XL600 Millipore Express SHR single layer 0.1 um 9/16 in. HB/HB with Bell
Catalog No. NC0333625