Learn More
MSC Reynolon 5044 Shrink Film, 14 x 1500 ft, 1 Case
Catalog No. 501919376
Encompass
Reynolon shrink films are designed to enhance product appeal with their strength, durability, sparkling clarity, and high gloss, making them suitable for various packaging needs.
  • Soft touch
  • Medium biaxial shrinkage
  • Suitable for non-food packaging
  • Industry standard general purpose film
  • Outstanding sparkle, gloss, and wrinkle-free shrinkage
  • High internal tear resistance
  • Good memory
  • Center-folded film
Catalog No. 50-191-9376 Supplier MSC Supplier No. 39589890
Reynolon shrink films are designed to enhance product appeal with their strength, durability, sparkling clarity, and high gloss, making them suitable for various packaging needs.
  • Soft touch
  • Medium biaxial shrinkage
  • Suitable for non-food packaging
  • Industry standard general purpose film
  • Outstanding sparkle, gloss, and wrinkle-free shrinkage
  • High internal tear resistance
  • Good memory
  • Center-folded film