Learn More
MSC Shrink Film, 16 in Width, 2500 ft
Catalog No. 501918471
Encompass
This polyolefin shrink film is designed for packaging various items, offering excellent performance across different equipment types. It provides a protective and visually appealing finish for a wide range of applications.
  • Excellent performance on manual, semi-automatic, and automatic equipment
  • Multi-layer, biaxially oriented film requires no dwell time
  • High gloss appearance and excellent clarity
  • Puncture resistant
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded film
Catalog No. 50-191-8471 Supplier MSC Supplier No. 39580618
This polyolefin shrink film is designed for packaging various items, offering excellent performance across different equipment types. It provides a protective and visually appealing finish for a wide range of applications.
  • Excellent performance on manual, semi-automatic, and automatic equipment
  • Multi-layer, biaxially oriented film requires no dwell time
  • High gloss appearance and excellent clarity
  • Puncture resistant
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded film