Learn More
MSC Reynolon Polyolefin Shrink Film, 16 in, 75 Gauge, 1 Roll
Catalog No. 501919372
Encompass
This multi-layer, biaxially oriented Polyolefin Shrink Film offers excellent performance on manual, semi-automatic, and automatic equipment. It provides a high gloss appearance, excellent clarity, and is puncture resistant.
  • Requires no dwell time
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded
Catalog No. 50-191-9372 Supplier MSC Supplier No. 39589858
This multi-layer, biaxially oriented Polyolefin Shrink Film offers excellent performance on manual, semi-automatic, and automatic equipment. It provides a high gloss appearance, excellent clarity, and is puncture resistant.
  • Requires no dwell time
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded