Learn More
MSC MSC Industrial Shrink Wrap, 8 in, 60 Gauge, 1 Roll
Catalog No. 501919364
Encompass
This multi-layer, biaxially oriented shrink film is designed for efficient packaging across manual, semi-automatic, and automatic equipment. It offers high gloss and clarity, superior puncture resistance, and is suitable for various items, including those stored in cold environments.
  • Made in USA
  • Excellent performance on all types of manual, semi-automatic and automatic equipment
  • Requires no dwell time
  • High gloss appearance and excellent clarity
  • Puncture resistant
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded film
Catalog No. 50-191-9364 Supplier MSC Supplier No. 39589775
This multi-layer, biaxially oriented shrink film is designed for efficient packaging across manual, semi-automatic, and automatic equipment. It offers high gloss and clarity, superior puncture resistance, and is suitable for various items, including those stored in cold environments.
  • Made in USA
  • Excellent performance on all types of manual, semi-automatic and automatic equipment
  • Requires no dwell time
  • High gloss appearance and excellent clarity
  • Puncture resistant
  • Works well on irregularly shaped items
  • Suitable for cold storage applications
  • Meets FDA requirements for food contact
  • Approved for mailings by the U.S. Postal Service
  • Center-folded film