Learn More
Grainger Baileigh Industrial Metal Shrinker Stretcher, 16 Gauge Steel Capacity, 6 in. Throat Depth, Foot Pedal Operated, 1 ea.
Catalog No. 502784651
Encompass_Preferred
This industrial metal shrinker stretcher is designed for precision metalworking and fabrication, featuring a robust steel frame and a durable cast iron stand for reliable performance. The unit utilizes manual, foot pedal-activated controls to provide hands-free operation, allowing users to guide materials effectively while maintaining consistent forming precision.
  • Capacity for 16 gauge mild steel and 14 gauge aluminum
  • 6 in. throat depth
  • Includes two quick-change jaw sets for shrinking and stretching
  • Rounded jaws to minimize material marking
  • Adjustable forming depth and pressure
  • Integrated wrench holder for convenience
  • Rolling casters for mobility
  • Fast tooling changes
Catalog No. 50-278-4651 Supplier Grainger Supplier No. 18F322
This industrial metal shrinker stretcher is designed for precision metalworking and fabrication, featuring a robust steel frame and a durable cast iron stand for reliable performance. The unit utilizes manual, foot pedal-activated controls to provide hands-free operation, allowing users to guide materials effectively while maintaining consistent forming precision.
  • Capacity for 16 gauge mild steel and 14 gauge aluminum
  • 6 in. throat depth
  • Includes two quick-change jaw sets for shrinking and stretching
  • Rounded jaws to minimize material marking
  • Adjustable forming depth and pressure
  • Integrated wrench holder for convenience
  • Rolling casters for mobility
  • Fast tooling changes