Learn More
Grainger Sharps Container 2 Gal. Rotor Lid PK5
Catalog No. NC9754275
Encompass_Preferred
Sharps Disposal Box,2-gal,Clear Rotor Lid,5/pack,containers with Rotor Lids have a stepped unwinder to let you remove needles safely. Also include both temporary and permanent lid locks,lid designs keep hands away from discarded sharps
Catalog No. NC9754275 Supplier Grainger Supplier No. 3UTE7
Sharps Disposal Box,2-gal,Clear Rotor Lid,5/pack,containers with Rotor Lids have a stepped unwinder to let you remove needles safely. Also include both temporary and permanent lid locks,lid designs keep hands away from discarded sharps