Learn More
Grainger Sharps Container Red Round 5 Qt PK5
Catalog No. NC9795564
Encompass_Preferred
Sharps Disposal Box, 5-qt, Red Rotor Lid, 5/pack, Specifications:Puncture-resistant, rigid plastic contains sharps securely. Can be autoclaved or incinerated, Sharp's Disposal System (No. 55535)
This item is not returnable. View return policy
Catalog No. NC9795564 Supplier Grainger Supplier No. 3UTE6
Sharps Disposal Box, 5-qt, Red Rotor Lid, 5/pack, Specifications:Puncture-resistant, rigid plastic contains sharps securely. Can be autoclaved or incinerated, Sharp's Disposal System (No. 55535)