Learn More
Andwin Scientific SHRPS CONTNR SYS RED MED 24/CS
SDP
Catalog No. NC0019804
Encompass_Preferred
Stackable Sharps Container Safety System Medium 3.8L. For safe and effective containment and disposal of contaminated needles syringes and other sharps. Puncture-resistant and autoclave/incinerator-safe. Features transparent locking lid and needle rem
Catalog No. NC0019804 Supplier Andwin Scientific Supplier No. 19001003
Stackable Sharps Container Safety System Medium 3.8L. For safe and effective containment and disposal of contaminated needles syringes and other sharps. Puncture-resistant and autoclave/incinerator-safe. Features transparent locking lid and needle rem