Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich SHT-263 Solarpur™

Catalog No. 9025001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-250-01G 1 g
90-250-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-250-01G Supplier Sigma Aldrich Supplier No. 9025001G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SHT-263 Solarpur™

Specifications

CAS 207739-72-8
Melting Point 247°C
Molecular Formula C81H68N4O8
Linear Formula C81H68N4O8
MDL Number MFCD12022511
Quantity 1 g
Synonym N2,N2,N2,N2,N7,N7,N7,N7-octakis(4-methoxyphenyl)-9,9 -spirobi[9 H-fluorene]-2,2 ,7,7 -tetramine; Spiro-MeOTAD; Spiro-OMeTAD
Molecular Weight (g/mol) 1225.43
Percent Purity ≥99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.