Learn More
Abcam SHU9119, Melanocortin 3 and 4 (MC3 and MC4) receptor antagonist, 1MG
Catalog No. 502706845
Encompass_Preferred
MW 1074.25 g/mol, Purity >95%. Potent melanocortin 3 and 4 (MC₃ and MC₄) receptor antagonist (IC₅₀ values are 0.23 and 0.06 nM, respectively). Also shows MC₅ partial agonist activity (EC₅₀ = 0.12 nM). Dose-dependently increases food intake following central administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6845 Supplier Abcam Supplier No. AB1411641MG
MW 1074.25 g/mol, Purity >95%. Potent melanocortin 3 and 4 (MC₃ and MC₄) receptor antagonist (IC₅₀ values are 0.23 and 0.06 nM, respectively). Also shows MC₅ partial agonist activity (EC₅₀ = 0.12 nM). Dose-dependently increases food intake following central administration.
The product is subject to the following: Abcam Restricted Use Statement