Learn More
Electron Microscopy Sciences The most up-to-date technology for slide drying method. Adjustable shelf accepts an assortment of slide racks with and/or without coverslipper adapters. Adjustable air flow - Digital programming - Microprocessor controlled.
SDP
Catalog No. NC3593866
Encompass_Preferred
This is the most up-to-date technology apply to slide drying method. This new design allows users to increase their productivity while maintaining super results. Adjustable shelf accepts an assortment of slide racks with and/or without coverslipper adapters. Temperature from ambient to 70 degree C +/- 1 degree C. Adjustable air flow and 51 CFM of air flow. Digital programming, microprocessor controlled, and recessed 9 W fluorescent lighting. Four 3,200 rpm fans.
Catalog No. NC3593866 Supplier Electron Microscopy Sciences Supplier No. 6322410
Add to Cart
Add to Cart
Request a Quote
This is the most up-to-date technology apply to slide drying method. This new design allows users to increase their productivity while maintaining super results. Adjustable shelf accepts an assortment of slide racks with and/or without coverslipper adapters. Temperature from ambient to 70 degree C +/- 1 degree C. Adjustable air flow and 51 CFM of air flow. Digital programming, microprocessor controlled, and recessed 9 W fluorescent lighting. Four 3,200 rpm fans.