Learn More
Electron Microscopy Sciences SHUR/Dry Microscope Slide Dryer III, 220V
SDP
Catalog No. 501901639
Encompass_Preferred
This is the most up-to-date technology apply to slide drying method. This new design allows users to increase their productivity while maintaining super results. Adjustable shelf accepts an assortment of slide racks with and/or without coverslipper adapters. Temperature from ambient to 70 degree C +/- 1 degree C. Adjustable air flow and 51 CFM of air flow. Digital programming, microprocessor controlled, and recessed 9 W fluorescent lighting. Four 3,200 rpm fans.
Catalog No. 50-190-1639 Supplier Electron Microscopy Sciences Supplier No. 6322420
Add to Cart
Add to Cart
Request a Quote
This is the most up-to-date technology apply to slide drying method. This new design allows users to increase their productivity while maintaining super results. Adjustable shelf accepts an assortment of slide racks with and/or without coverslipper adapters. Temperature from ambient to 70 degree C +/- 1 degree C. Adjustable air flow and 51 CFM of air flow. Digital programming, microprocessor controlled, and recessed 9 W fluorescent lighting. Four 3,200 rpm fans.