Learn More
Chemglass Life Sciences Adapter, 0-20mm, Vacuum Shut Off, Airfree, Schlenk
SDP
Catalog No. AF030006
Encompass_Preferred
Adapter, 0-20mm, Vacuum Shut Off, Airfree, Schlenk. A 0-20mm Chem-Vac™ Chem-Cap valve having a #20 o-ring joint for connection to the AF-0300-04 trap assembly and a 1" OD lower arm for connection to the rubber vacuum tubing. Adapter is supplied complete with a #214 Viton o-ring for use with the #20 o-ring joint.
Catalog No. AF030006 Supplier Chemglass Life Sciences Supplier No. AF030006
Adapter, 0-20mm, Vacuum Shut Off, Airfree, Schlenk. A 0-20mm Chem-Vac™ Chem-Cap valve having a #20 o-ring joint for connection to the AF-0300-04 trap assembly and a 1" OD lower arm for connection to the rubber vacuum tubing. Adapter is supplied complete with a #214 Viton o-ring for use with the #20 o-ring joint.