Learn More
Analytical Sales & Services Shut-Off Valve for 1/8" OD Tubing, Tefzel™ (ETFE)
Catalog No. 5000807078
Encompass
Shut-Off Valve for 1/8" OD Tubing\n• Tefzel™ (ETFE)\n• .040" Thru-Hole\n• Includes 1/8" Fittings\n\nQty/Pkg: EACH
Catalog No. 50-008-07078 Supplier Analytical Sales & Services Supplier No. 59783
Shut-Off Valve for 1/8" OD Tubing\n• Tefzel™ (ETFE)\n• .040" Thru-Hole\n• Includes 1/8" Fittings\n\nQty/Pkg: EACH