Learn More
Julabo USA Inc Each, shut off vavle, G 1 1/4" male/female fittings
SDP
Catalog No. 507534246
Encompass_Preferred
Bath Accessories, Julabo, Shut-off valve G 1 1/4″ male/female fittings. For use with 6 bar pump FL chillers: FL7006, FLW7006, FL11006, FLW11006, FL20006, FLW20006.
Catalog No. 50-753-4246 Supplier Julabo USA Inc Supplier No. 8970458
Bath Accessories, Julabo, Shut-off valve G 1 1/4″ male/female fittings. For use with 6 bar pump FL chillers: FL7006, FLW7006, FL11006, FLW11006, FL20006, FLW20006.