Learn More
Julabo USA Inc Each, shut off vavle, G 3/4" male/female fittings
SDP
Catalog No. 507534243
Encompass_Preferred
Bath Accessories, Julabo, Shut-off valve G 3/4″ male/female. For use with 3 bar pump FL chillers: FL1203, FL1703, FLW1703, FL2503, FLW2503, FL4003, FLW4003.
Catalog No. 50-753-4243 Supplier Julabo USA Inc Supplier No. 8970454
Bath Accessories, Julabo, Shut-off valve G 3/4″ male/female. For use with 3 bar pump FL chillers: FL1203, FL1703, FLW1703, FL2503, FLW2503, FL4003, FLW4003.