Learn More
Grainger Dormont Quick Disconnect Gas Shutoff Valve, Brass, 1 in, 1 ea
Catalog No. 502778674
Encompass_Preferred
This 1 in. quick disconnect gas shutoff valve is engineered for the foodservice industry, designed to provide safe and efficient gas flow management for commercial cooking equipment. The fitting features a push-to-connect mechanism and includes an integrated thermal shutoff for enhanced safety. Its robust construction ensures reliable operation in demanding kitchen environments.
  • Features a push-to-connect quick disconnect with thermal shutoff
  • Includes a carbon steel safety sleeve that prevents disconnection until the gas valve is closed
  • Designed so the valve cannot be opened until the gas connector is securely attached
  • Constructed from durable brass
  • Suitable for gas service
Catalog No. 50-277-8674 Supplier Grainger Supplier No. 18D844
This 1 in. quick disconnect gas shutoff valve is engineered for the foodservice industry, designed to provide safe and efficient gas flow management for commercial cooking equipment. The fitting features a push-to-connect mechanism and includes an integrated thermal shutoff for enhanced safety. Its robust construction ensures reliable operation in demanding kitchen environments.
  • Features a push-to-connect quick disconnect with thermal shutoff
  • Includes a carbon steel safety sleeve that prevents disconnection until the gas valve is closed
  • Designed so the valve cannot be opened until the gas connector is securely attached
  • Constructed from durable brass
  • Suitable for gas service