Learn More
IDEX Health & Science Upchurch SHUTOFF VLV .040IN THRU TEFZEL
Catalog No. NC1920882
Encompass_Preferred
Idex Shut-off Valves, Diameter Outer: 0.317 cm, 0.125 in., Tefzel (ETFE), Pressure: 600 psig, Shut-Off Valve, Includes: 1/4-28 fittings for tubing
Catalog No. NC1920882 Supplier IDEX Health & Science Upchurch Supplier No. P721
Idex Shut-off Valves, Diameter Outer: 0.317 cm, 0.125 in., Tefzel (ETFE), Pressure: 600 psig, Shut-Off Valve, Includes: 1/4-28 fittings for tubing