Learn More
MSC Fantech Shutter Mount Exhaust Fan, 24 in., 1/4 HP, 1 Phase, 1 ea.
Catalog No. 5000507064
Encompass
This 24 in. shutter mount exhaust fan is designed for efficient ventilation in diverse environments, including warehouses, factories, workshops, and agricultural buildings. This unit features a direct drive motor and is engineered for straightforward, ready-to-install mounting using pre-punched holes in the shutter frame.
  • UL 705 standard listed for safety compliance
  • Totally enclosed motor with speed control capabilities
  • Heavy-duty guards with a gray polyester coating to resist corrosion
  • OSHA-compliant guards for enhanced operational safety
  • Aluminum blades for durable performance
  • Shipped ready to install for immediate setup
Catalog No. 50-005-07064 Supplier MSC Supplier No. 74077611
This 24 in. shutter mount exhaust fan is designed for efficient ventilation in diverse environments, including warehouses, factories, workshops, and agricultural buildings. This unit features a direct drive motor and is engineered for straightforward, ready-to-install mounting using pre-punched holes in the shutter frame.
  • UL 705 standard listed for safety compliance
  • Totally enclosed motor with speed control capabilities
  • Heavy-duty guards with a gray polyester coating to resist corrosion
  • OSHA-compliant guards for enhanced operational safety
  • Aluminum blades for durable performance
  • Shipped ready to install for immediate setup