Learn More
Azzota Corp Shower Safety Station W/ Eyewash Stainless Steel Bowl, Abs Body And Bowl, Shower Height: 88 Inch To 91 Inch, Shower Head Diameter: 8 Inch, Eye Wash Height: 37 Inch To 41 Inch
Catalog No. 501612838
Encompass_Preferred
Shower Safety Station w/ Eyewash Stainless Steel Bowl, ABS body and Bowl..Meet US Z358.1-2004 standard, Shower height: 88 inch to 91 inch, Shower Head Diameter: 8 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 13 inch, Material: 304...
Catalog No. 50-161-2838 Supplier Azzota Corp Supplier No. EW8837PT
Shower Safety Station w/ Eyewash Stainless Steel Bowl, ABS body and Bowl..Meet US Z358.1-2004 standard, Shower height: 88 inch to 91 inch, Shower Head Diameter: 8 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 13 inch, Material: 304...