Learn More
Azzota Corp Shower Safety Station W/Eyewash Bowl, Shower Height: 88 Inch To 91 Inch, Shower Head Diameter: 8 Inch, Eye Wash Height: 37 Inch To 41 Inch, Eye Wash Bowl Diameter: 13 Inch, M...
Catalog No. 501612839
Encompass_Preferred
Shower Safety Station w/ Eyewash Stainless Steel Bowl, Stainless Steel body and Bowl..Meet US Z358.1-2004 standard, Shower height: 88 inch to 91 inch, Shower Head Diameter: 8 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 13 inch, M...
Catalog No. 50-161-2839 Supplier Azzota Corp Supplier No. EW8837SA
Shower Safety Station w/ Eyewash Stainless Steel Bowl, Stainless Steel body and Bowl..Meet US Z358.1-2004 standard, Shower height: 88 inch to 91 inch, Shower Head Diameter: 8 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 13 inch, M...