Learn More
Azzota Corp Shower Safety Station W/ Eyewash Bowl, Ss Body/Bowl, Height: 82-91", Head Diameter: 10", Ew Height: 37-41", Bowl Diameter: 11", Material: 304 Ss, Timing: 1 Second, Volume: 0.2L-0.3L/S, Volume: 2L-3L/S
Catalog No. 501612840
Encompass_Preferred
Shower Safety Station w/ Eyewash Stainless Steel Bowl, Stainless Steel body and Bowl, Meet US Z358.1-2004 standard, Shower height: 82 inch to 91 inch, Shower Head Diameter: 10 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 11 inch, Material: 304 Stainless Steel, Turn on timing: 1 second, Spray volume: 0.2L-0.3L/S, Shower Volume: 2L-3L/S
Catalog No. 50-161-2840 Supplier Azzota Corp Supplier No. EW8837SS
Shower Safety Station w/ Eyewash Stainless Steel Bowl, Stainless Steel body and Bowl, Meet US Z358.1-2004 standard, Shower height: 82 inch to 91 inch, Shower Head Diameter: 10 inch, Eye wash height: 37 inch to 41 inch, Eye wash Bowl diameter: 11 inch, Material: 304 Stainless Steel, Turn on timing: 1 second, Spray volume: 0.2L-0.3L/S, Shower Volume: 2L-3L/S