Learn More
AdipoGen Shz-1 (1 mg)
SDP
Catalog No. 502053877
Encompass_Preferred
Chemical. CAS: 326886-05-9. Formula: C13H11BrN2O3S. MW: 355.2. Enhancer of myocardial regenerative repair by stem cells. Potent inducer of Nkx2.5 and a subset of other cardiac markers, including myocardin, and sarcomeric-tropomyosin (SalphaTM) in a variety of different stem/progenitor cells.
Catalog No. 50-205-3877 Supplier AdipoGen Supplier No. AGCR13502M001
Chemical. CAS: 326886-05-9. Formula: C13H11BrN2O3S. MW: 355.2. Enhancer of myocardial regenerative repair by stem cells. Potent inducer of Nkx2.5 and a subset of other cardiac markers, including myocardin, and sarcomeric-tropomyosin (SalphaTM) in a variety of different stem/progenitor cells.