Learn More
Medchemexpress LLC HY-129603 5mg Medchemexpress, SI-109 CAS:2429877-30-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1296035MG
Encompass_Preferred
Medchemexpress, HY-129603 5mg SI-109 CAS:2429877-30-3 SI-109 is a potent STAT3 SH2 domain inhibitor (Ki=9 nM) with antitumor activity. SI-109 effectively inhibits the transcriptional activity of STAT3 (IC50=3 μM). SI-109 and an analog of CRBN ligand lenalidomide have been used to design PROTAC STAT3 degrader SD-36. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1215