Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Si/1G-44H2SO4/Si SPE Tube

Catalog No. 111017090 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7090 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7090 Supplier MilliporeSigma™ Supelco™ Supplier No. 57145U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

TRUSTED_SUSTAINABILITY

Specifications

Column Phase Si/1 G-44H2SO4/Si
Column Type SPE
For Use With (Application) Food and Beverages
Type Solid Phase Extraction Column
Volume (Metric) 12 mL
Material Polypropylene
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.