Learn More
TARGETMOL CHEMICALS INC SI-2 5MG
SDP
Catalog No. 502580493
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SI-2 is an inhibitor (SMI) of steroid receptor coactivator-3 (SRC-3 or AIB1). SI-2 can selectively reduce the transcriptional activities and the protein concentrations of SRC-3 in cells and selectively induce breast cancer cell death with IC50 values in the low nanomolar range (3-20 nM) but not affect normal cell viability. purity: 98%
Catalog No. 50-258-0493 Supplier TARGETMOL CHEMICALS INC Supplier No. T287735MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SI-2 is an inhibitor (SMI) of steroid receptor coactivator-3 (SRC-3 or AIB1). SI-2 can selectively reduce the transcriptional activities and the protein concentrations of SRC-3 in cells and selectively induce breast cancer cell death with IC50 values in the low nanomolar range (3-20 nM) but not affect normal cell viability. purity: 98%