Learn More
Aviva Systems Biology Corp SI Antibody - middle region (ARP44831_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44831_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SI. Alternate names: MGC131621, MGC131622ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58909