Learn More
Hach Company OriFlo™ Filter Apparatus
Supplier:  Hach Company 4966000
Encompass
Designed for rapid, qualitative filtration of particulate matter from aqueous samples using a 25 mm glass fiber filter or other medium with porosity greater than or equal to 1 micron.
Catalog No. 50-199-7115