Learn More
Hach Company Safety Shield
Supplier:  Hach Company 5003000
Encompass
For use with Hach Digesdhal Apparatus or COD Reactor. Order 5003000 for COD Reactor, Polycarbonate Shield 15"Height x 16" Width. Order 5004000 for Hach Digesdhal, Polycarbonate Shield 30" Height x 16" Width, heavy 2.223 cm thick (0.875") urethane coated. Heavy base resists slipping on countertops and lab benches. Shield formed in a parabolic curve.
Catalog No. 50-199-7117