Learn More
LifeSensors Ubiquitin-K63R 1mg
SDP
Catalog No. NC3710504
Encompass_Preferred
Ubiquitin-K63R is recombinant ubiquitin with a single substitution of arginine for lysine at position 63. Ubiquitin is a small polypeptide that can be conjugated via its C-terminus to amine groups of lysine residue on target proteins. This conjunction is referred to as monoubiquitylation. Additional ubiquitin moieties can subsequently be conjugated to this initial ubiquitin, utilizing any one (of seven) lysine residues on the surface of ubiquitin. The formation of these ubiquitin chains is referred to as polyubiquitylation.
Catalog No. NC3710504 Supplier LifeSensors Supplier No. SI02181000
Ubiquitin-K63R is recombinant ubiquitin with a single substitution of arginine for lysine at position 63. Ubiquitin is a small polypeptide that can be conjugated via its C-terminus to amine groups of lysine residue on target proteins. This conjunction is referred to as monoubiquitylation. Additional ubiquitin moieties can subsequently be conjugated to this initial ubiquitin, utilizing any one (of seven) lysine residues on the surface of ubiquitin. The formation of these ubiquitin chains is referred to as polyubiquitylation.