Learn More
Bioworld SIAB, 100 mg
Catalog No. 50199263
Encompass_Preferred
SIAB, N-Succinimidyl (4-iodoacetyl) AminobenzoatePrimary amine and sulfhydryl reactiveEnzyme-antibody conjugationReferences: 1.) Weltman, J.K., et al. (1983) Bio. Techniques 1, 148-152.M.W. = 402.15
Catalog No. 50-199-263 Supplier Bioworld Supplier No. 218100151
SIAB, N-Succinimidyl (4-iodoacetyl) AminobenzoatePrimary amine and sulfhydryl reactiveEnzyme-antibody conjugationReferences: 1.) Weltman, J.K., et al. (1983) Bio. Techniques 1, 148-152.M.W. = 402.15