Learn More
LECTENZ BIO INC SiaFind Alpha 2,6-Specific Reagent, Biotinylated Kit
SDP
Catalog No. NC3401858
Encompass_Preferred
The SiaFind Alpha 2,6-Specific Reagent, Biotinylated Kit contains a recombinant protein engineered from Polyporus squamosus lectin (PSL). This highly purified affinity reagent is designed for sensitive, robust, and specific detection of Siaα2,6Gal commonly found in glycoconjugates (glycoproteins, glycolipids, and oligo- or polysaccharides). Applications include use as a primary probe in ELISA, Western blot, and immunohistochemistry. It is not recommended for flow cytometry.
Catalog No. NC3401858 Supplier LECTENZ BIO INC Supplier No. SK2601B
The SiaFind Alpha 2,6-Specific Reagent, Biotinylated Kit contains a recombinant protein engineered from Polyporus squamosus lectin (PSL). This highly purified affinity reagent is designed for sensitive, robust, and specific detection of Siaα2,6Gal commonly found in glycoconjugates (glycoproteins, glycolipids, and oligo- or polysaccharides). Applications include use as a primary probe in ELISA, Western blot, and immunohistochemistry. It is not recommended for flow cytometry.