Learn More
LECTENZ BIO INC SiaFind Pan-Specific Lectenz SureLight 488 Kit
SDP
Catalog No. NC2332647
Encompass_Preferred
The SiaFind Pan-Specific Lectenz SureLight 488 Kit contains a sialic acid affinity reagent for the detection, separation, or enrichment of sialoglycans terminating in Sia-alpha-2,3-Gal, Sia-alpha-2,6-Gal, and Sia-alpha-2,8-Sia commonly found in glycoconjugates (glycoproteins, glycolipids, and oligo- or polysaccharides). It has high affinity and specificity towards sialoglycans in a linkage independent manner (pan-specific). LECTENZ BIO mfg# SK0501F
Catalog No. NC2332647 Supplier LECTENZ BIO INC Supplier No. SK0501F
The SiaFind Pan-Specific Lectenz SureLight 488 Kit contains a sialic acid affinity reagent for the detection, separation, or enrichment of sialoglycans terminating in Sia-alpha-2,3-Gal, Sia-alpha-2,6-Gal, and Sia-alpha-2,8-Sia commonly found in glycoconjugates (glycoproteins, glycolipids, and oligo- or polysaccharides). It has high affinity and specificity towards sialoglycans in a linkage independent manner (pan-specific). LECTENZ BIO mfg# SK0501F