Learn More
Medchemexpress LLC Siais164018 hydrochloride | 00-00-0 | 98.1% | C43H49Cl2N10O7P | 10MG
SDP
Catalog No. 5000258223
Encompass_Preferred
SIAIS164018 hydrochloride is a PROTAC-based degrader that targets ALK and EGFR, demonstrating potent cellular activity to inhibit cancer cell migration, induce G1 cell cycle arrest, and promote apoptosis. Reported activity includes low-nanomolar potency in relevant assays. Supplied as a purified research compound for biochemical and cellular studies.
  • Potent ALK and EGFR degradation in cellular assays.
  • Inhibits cancer cell migration and invasion.
  • Induces G1 cell cycle arrest and apoptosis.
  • High reported purity and defined molecular properties.
  • Supplied in small-mass research quantities for in vitro studies.
Catalog No. 50-002-58223 Supplier Medchemexpress LLC Supplier No. HY147219A10MG
SIAIS164018 hydrochloride is a PROTAC-based degrader that targets ALK and EGFR, demonstrating potent cellular activity to inhibit cancer cell migration, induce G1 cell cycle arrest, and promote apoptosis. Reported activity includes low-nanomolar potency in relevant assays. Supplied as a purified research compound for biochemical and cellular studies.
  • Potent ALK and EGFR degradation in cellular assays.
  • Inhibits cancer cell migration and invasion.
  • Induces G1 cell cycle arrest and apoptosis.
  • High reported purity and defined molecular properties.
  • Supplied in small-mass research quantities for in vitro studies.