Learn More
GENOVIS INC SialEXO, SialEXO, 2000 units, Origin: E. Coli
Catalog No. NC1494512
Encompass
SialEXO is a mixture of sialidases hydrolyzing a2-3, a2-6 and a2-8 linked sialic acids. The SialEXO mix removes all sialic acids from N- and O-linked glycans on native (non-denatured) proteins and peptides. SialEXO is available as lyophilized powder for desialylation of 2 mg of sialylated glycoproteins.
This item is not returnable. View return policy
Catalog No. NC1494512 Supplier GENOVIS INC Supplier No. G1SM1020
SialEXO is a mixture of sialidases hydrolyzing a2-3, a2-6 and a2-8 linked sialic acids. The SialEXO mix removes all sialic acids from N- and O-linked glycans on native (non-denatured) proteins and peptides. SialEXO is available as lyophilized powder for desialylation of 2 mg of sialylated glycoproteins.