Learn More
Sigma Aldrich Fine Chemicals Biosciences Sialic Acid Aldolase from
Catalog No. 502906536
Encompass
Sialic acid aldolases or N-acetylneuraminate lyases catalyze the reversible aldol cleavage of N-acetylneuraminic acid to form pyruvate and N-acetyl-D-mannosamine. In nature N-acetylneuraminate lyase mainly occurs in pathogens.
Catalog No. 50-290-6536 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S182610UN
Sialic acid aldolases or N-acetylneuraminate lyases catalyze the reversible aldol cleavage of N-acetylneuraminic acid to form pyruvate and N-acetyl-D-mannosamine. In nature N-acetylneuraminate lyase mainly occurs in pathogens.