Learn More
Sigma Aldrich Fine Chemicals Biosciences Sialic Acid Assay Kit suff
Catalog No. 502904315
Encompass
Sialic Acid is a general name for nine carbon acidic sugars with N- or O-substituted derivatives. The most common member of these sugars is N-acetylneuraminic acid (NANA). They are associated with carbohydrates and are localized on cell membranes. Sialic acid is widely distributed throughout mammalian tissues and fluids including serum. Sialylated oligosaccharides have been shown to exhibit antiviral properties and are also known to influence blood coagulation and cholesterol levels. The sialic acid level in body fluids is also an important marker for diagnosing cancer.
Catalog No. 50-290-4315 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3141KT
Sialic Acid is a general name for nine carbon acidic sugars with N- or O-substituted derivatives. The most common member of these sugars is N-acetylneuraminic acid (NANA). They are associated with carbohydrates and are localized on cell membranes. Sialic acid is widely distributed throughout mammalian tissues and fluids including serum. Sialylated oligosaccharides have been shown to exhibit antiviral properties and are also known to influence blood coagulation and cholesterol levels. The sialic acid level in body fluids is also an important marker for diagnosing cancer.