Learn More
Takara Bio SIALIC ACID FLUORESCENCE LABEL
SDP
Catalog No. 50443829
Encompass
Labeling kit; Sialic Acid Fluorescence Labeling Kit; Takara; Storage: Store at -20 deg.C, DMB Solution in dark; DMB Solution, 2x1mL, Coupling Solution (2-mercaptoethanol, sodium hydrosulfite, acetic acid), 2x5mL, Neu5Ac (100uL), 500uL
Catalog No. 50-443-829 Supplier Takara Bio Supplier No. 4400
Labeling kit; Sialic Acid Fluorescence Labeling Kit; Takara; Storage: Store at -20 deg.C, DMB Solution in dark; DMB Solution, 2x1mL, Coupling Solution (2-mercaptoethanol, sodium hydrosulfite, acetic acid), 2x5mL, Neu5Ac (100uL), 500uL