Learn More
Sigma Aldrich Fine Chemicals Biosciences Sialic Acid Quantitation K
Catalog No. 502907958
Encompass
The Sialic Acid Quantitation Kit provides a rapid and accurate determination of total N-acetylneuraminic acid (NANA sialic acid). NANA is released from glycoconjugates using neuraminidase. The kit uses alpha(2-3 6 8 9) neuraminidase to cleave all NANA linkages including alpha(2-8) and alpha(2-9) linkages as well as branched NANA for the most accurate determination of total sialic acid content.
Catalog No. 50-290-7958 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SIALICQ1KT
The Sialic Acid Quantitation Kit provides a rapid and accurate determination of total N-acetylneuraminic acid (NANA sialic acid). NANA is released from glycoconjugates using neuraminidase. The kit uses alpha(2-3 6 8 9) neuraminidase to cleave all NANA linkages including alpha(2-8) and alpha(2-9) linkages as well as branched NANA for the most accurate determination of total sialic acid content.