Learn More
Agilent Technologies AdvanceBio total sialic acid quantitation kit, 48/pk
Catalog No. NC1514271
Encompass
Rapid quantitation of total sialic acid by fluorescence or absorbance detection, either free or released from intact glycoproteins. Automatable, 96-well microplate format with fluorescence detection. Broad range of detection, 40 pmol - 1,000 pmol. Sample digestion, conversion, detection and quantitation may be performed in a single well for fast and simple processing. Enzymatic cleavage allows rapid analysis (~75 minutes or less) with minimal, degradation of sialic acid.
Catalog No. NC1514271 Supplier Agilent Technologies Supplier No. GS48SAQ
Rapid quantitation of total sialic acid by fluorescence or absorbance detection, either free or released from intact glycoproteins. Automatable, 96-well microplate format with fluorescence detection. Broad range of detection, 40 pmol - 1,000 pmol. Sample digestion, conversion, detection and quantitation may be performed in a single well for fast and simple processing. Enzymatic cleavage allows rapid analysis (~75 minutes or less) with minimal, degradation of sialic acid.