Learn More
QA Bio Inc Ludger Sialic acid reference panel
Supplier:  QA Bio Inc CMSRP01
Encompass
The Sialic Acid Reference Panel contains Neu5Ac; Neu5Gc; Neu5,7Ac2; Neu5,Gc9Ac; Neu5,9Ac2; Neu5,8Ac2 and Neu5,7,(8),9Ac3Gc. • Also supplied as a component of our LudgerTag DMB sialic acid release and labeling kit (LT-KDMB-A1). • Sources: Bovine submaxillary mucin • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Amount: 1.25 nmols
Catalog No. 50-223-6583